missing translation for 'onlineSavingsMsg'
Learn More

Chlorpyrifos, Restek

Catalog No. 06719844
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06719844 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06719844 Supplier Chromatographic Supplier No. RK 32212

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2921-88-2
Molecular Formula C9H11Cl3NO3PS
Molecular Weight (g/mol) 350.575
InChI Key SBPBAQFWLVIOKP-UHFFFAOYSA-N
PubChem CID 2730
ChEBI CHEBI:34631
IUPAC Name diethoxy-sulfanylidene-(3,5,6-trichloropyridin-2-yl)oxy-$l^{5}-phosphane
SMILES CCOP(=S)(OCC)OC1=NC(=C(C=C1Cl)Cl)Cl

Specifications

Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000ug/mL, Methanol
Chemical Name or Material Chlorpyrifos, Pesticide Mix