missing translation for 'onlineSavingsMsg'
Learn More

Chlorpropham 99.0+%, TCI America™

Catalog No. C25551G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25551G 1 g
C255525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25551G Supplier TCI America Supplier No. C25551G

Chemical Identifiers

CAS 101-21-3
Molecular Formula C10H12ClNO2
Molecular Weight (g/mol) 213.66
MDL Number MFCD00037108
InChI Key CWJSHJJYOPWUGX-UHFFFAOYSA-N
Synonym chlorpropham, chloropropham, chloro-ipc, isopropyl m-chlorocarbanilate, cipc, chlorprophame, elbanil, isopropyl n-3-chlorophenyl carbamate, metoxon, nexoval
PubChem CID 2728
ChEBI CHEBI:34630
IUPAC Name propan-2-yl N-(3-chlorophenyl)carbamate
SMILES CC(C)OC(=O)NC1=CC=CC(Cl)=C1

Specifications

Melting Point 42°C
Color White-Yellow
Quantity 1 g
UN Number 3077
Formula Weight 213.66
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Chlorpropham
EINECSNumber (3)-3217
RTECSNumber FD8050000
TSCA No