Learn More
Chlorpropamide
Increases insulin secretion | CAS: 94-20-2 | C10H13ClN2O3S | 276.735 g/mol
$147.04 - $480.16
Chemical Identifiers
| CAS | 94-20-2 |
|---|---|
| Molecular Formula | C10H13ClN2O3S |
| Molecular Weight (g/mol) | 276.735 |
| MDL Number | MFCD00079004 |
| InChI Key | RKWGIWYCVPQPMF-UHFFFAOYSA-N |
| Synonym | chlorpropamide, chloropropamide, diabinese, chlorpropamid, diabenese, glucamide, meldian, chlorodiabina, chloronase, diabeneza |
| PubChem CID | 2727 |
| ChEBI | CHEBI:3650 |
| IUPAC Name | 1-(4-chlorophenyl)sulfonyl-3-propylurea |
| SMILES | CCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6411014
|
Thermo Scientific Chemicals
J6411014 |
25 g |
N/A
|
|
|||||
|
AAJ6411022
|
Thermo Scientific Chemicals
J6411022 |
100 g |
N/A
|
|
|||||
Description
Chlorpropamide is used in the treatment of diabetes metilus type 2. It acts to increase the secretion of insulin and is not effective in patients who do not have pancreatic beta cell function.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorpropamide is used in the treatment of diabetes metilus type 2. It acts to increase the secretion of insulin and is not effective in patients who do not have pancreatic beta cell function.
Solubility
Soluble in ethanol (1:12), acetone (1:5), chloroform (1:9) and solutions of alkali hydroxides. Does not mix well with water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 94-20-2 | |
| 276.735 | |
| RKWGIWYCVPQPMF-UHFFFAOYSA-N | |
| 2727 | |
| 1-(4-chlorophenyl)sulfonyl-3-propylurea |
| C10H13ClN2O3S | |
| MFCD00079004 | |
| chlorpropamide, chloropropamide, diabinese, chlorpropamid, diabenese, glucamide, meldian, chlorodiabina, chloronase, diabeneza | |
| CHEBI:3650 | |
| CCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)Cl |
Specifications
| 94-20-2 | |
| C10H13ClN2O3S | |
| 25 g | |
| chlorpropamide, chloropropamide, diabinese, chlorpropamid, diabenese, glucamide, meldian, chlorodiabina, chloronase, diabeneza | |
| RKWGIWYCVPQPMF-UHFFFAOYSA-N | |
| 1-(4-chlorophenyl)sulfonyl-3-propylurea | |
| 2727 | |
| 276.74 | |
| Chlorpropamide |
| White | |
| MFCD00079004 | |
| 14,2186 | |
| Soluble in ethanol (1:12),acetone (1:5),chloroform (1:9) and solutions of alkali hydroxides. Does not mix well with water. | |
| CCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)Cl | |
| 276.735 | |
| CHEBI:3650 | |
| Powder |
Safety and Handling
GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.
P201-P202-P261-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P501c
H312+H332-H351
EINECSNumber : 202-314-5
RTECSNumber : YS6650000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only