missing translation for 'onlineSavingsMsg'
Learn More

Chlorpropamide

Catalog No. AAJ6411014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6411014 25 g
AAJ6411022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6411014 Supplier Thermo Scientific Chemicals Supplier No. J6411014

Increases insulin secretion | CAS: 94-20-2 | C10H13ClN2O3S | 276.735 g/mol

Chlorpropamide is used in the treatment of diabetes metilus type 2. It acts to increase the secretion of insulin and is not effective in patients who do not have pancreatic beta cell function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorpropamide is used in the treatment of diabetes metilus type 2. It acts to increase the secretion of insulin and is not effective in patients who do not have pancreatic beta cell function.

Solubility
Soluble in ethanol (1:12), acetone (1:5), chloroform (1:9) and solutions of alkali hydroxides. Does not mix well with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 94-20-2
Molecular Formula C10H13ClN2O3S
Molecular Weight (g/mol) 276.735
MDL Number MFCD00079004
InChI Key RKWGIWYCVPQPMF-UHFFFAOYSA-N
Synonym chlorpropamide, chloropropamide, diabinese, chlorpropamid, diabenese, glucamide, meldian, chlorodiabina, chloronase, diabeneza
PubChem CID 2727
ChEBI CHEBI:3650
IUPAC Name 1-(4-chlorophenyl)sulfonyl-3-propylurea
SMILES CCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)Cl

Specifications

Color White
Quantity 25 g
Merck Index 14,2186
Solubility Information Soluble in ethanol (1:12),acetone (1:5),chloroform (1:9) and solutions of alkali hydroxides. Does not mix well with water.
Formula Weight 276.74
Physical Form Powder
Chemical Name or Material Chlorpropamide
Hazard Category H312+H332-H351
Hazard Statement GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.
Precautionary Statement P201-P202-P261-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P501c
EINECSNumber 202-314-5
RTECSNumber YS6650000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only