missing translation for 'onlineSavingsMsg'
Learn More

Chlorotrimethylsilane, TMCS, EMD-Millipore

Catalog No. p-4782898
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1023330100 100 mL Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1023330100 Supplier Technidata Supplier No. 1.02333.0100
Only null left

Chemical Identifiers

CAS 75-77-4
Molecular Formula C3H9ClSi
Molecular Weight (g/mol) 108.64
InChI Key IJOOHPMOJXWVHK-UHFFFAOYSA-N
Synonym Trimethylchlorosilane, Trimethylsilyl chloride, TMCS
PubChem CID 6397
ChEBI CHEBI:85069
IUPAC Name chloro(trimethyl)silane
SMILES C[Si](C)(C)Cl

Specifications

Packaging Glass
Explosion Limit 1.5 to 46 %(V)
Ignition Point 420°C
Vapor Pressure 250 hPa (20°C)
Density Density: 0.86g/cm3 (20°C)
Quantity 100 mL
Formula Weight 108.64
Chemical Name or Material Chlorotrimethylsilane
Recommended Storage Store without restrictions