accessibility menu, dialog, popup

UserName

CAS RN 75-77-4

IUPAC Name: chlorotrimethylsilane
Synonyms: chlorotrimethylsilane
Molecular Weight (g/mol): 108.64
Molecular Formula: C3H9ClSi
InChi Key: IJOOHPMOJXWVHK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)Cl
Percent Purity 
  • (3)
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results