missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Chloromethylene)dimethyliminium Chloride 95.0+%, TCI America™
Supplier: TCI America C154525G
Specifications
| (Chloromethylene)dimethyliminium Chloride | |
| C3H7Cl2N | |
| 25 g | |
| arnold's reagent, vilsmeier reagent, chloromethylene dimethyliminium chloride, methanaminium, n-chloromethylene-n-methyl-, chloride, chloromethylene dimethylammonium chloride, n-chloromethylidene-n-methylmethanaminium chloride, chloromethylenedimethylammonium chloride, chloromethylidene dimethylazanium chloride, n-chloromethylene-n,n-dimethyl ammonium chloride | |
| C[N+](=CCl)C.[Cl-] | |
| 127.996 | |
| 128.00 | |
| Crystalline Powder |
| 3724-43-4 | |
| MFCD00011868 | |
| 2923 | |
| QQVDYSUDFZZPSU-UHFFFAOYSA-M | |
| chloromethylidene(dimethyl)azanium;chloride | |
| 77311 | |
| ≥95.0% (T) |
Chemical Identifiers
| 3724-43-4 | |
| 127.996 | |
| QQVDYSUDFZZPSU-UHFFFAOYSA-M | |
| 77311 | |
| C[N+](=CCl)C.[Cl-] |
| C3H7Cl2N | |
| MFCD00011868 | |
| arnold's reagent, vilsmeier reagent, chloromethylene dimethyliminium chloride, methanaminium, n-chloromethylene-n-methyl-, chloride, chloromethylene dimethylammonium chloride, n-chloromethylidene-n-methylmethanaminium chloride, chloromethylenedimethylammonium chloride, chloromethylidene dimethylazanium chloride, n-chloromethylene-n,n-dimethyl ammonium chloride | |
| chloromethylidene(dimethyl)azanium;chloride |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator