missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethylene)dimethyliminium Chloride 95.0+%, TCI America™

Catalog No. p-7125050
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C154525G 25 g
C1545250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C154525G Supplier TCI America Supplier No. C154525G
Only null left

Chemical Identifiers

CAS 3724-43-4
Molecular Formula C3H7Cl2N
Molecular Weight (g/mol) 127.996
MDL Number MFCD00011868
InChI Key QQVDYSUDFZZPSU-UHFFFAOYSA-M
Synonym arnold's reagent, vilsmeier reagent, chloromethylene dimethyliminium chloride, methanaminium, n-chloromethylene-n-methyl-, chloride, chloromethylene dimethylammonium chloride, n-chloromethylidene-n-methylmethanaminium chloride, chloromethylenedimethylammonium chloride, chloromethylidene dimethylazanium chloride, n-chloromethylene-n,n-dimethyl ammonium chloride
PubChem CID 77311
IUPAC Name chloromethylidene(dimethyl)azanium;chloride
SMILES C[N+](=CCl)C.[Cl-]

Specifications

Quantity 25 g
UN Number 2923
Formula Weight 128.00
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (Chloromethylene)dimethyliminium Chloride
TSCA Yes
Recommended Storage Refrigerator