missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Phenyl Sulfone 98.0+%, TCI America™

Supplier:  TCI America C15715G

 View more versions of this product

Catalog No. C15715G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Chloromethyl Phenyl Sulfone
53°C
130°C
MFCD00007551
chloromethyl phenyl sulfone, chloromethyl sulfonyl benzene, phenyl chloromethyl sulfone, chloromethylphenylsulfone, benzene, chloromethyl sulfonyl, chloromethylphenyl sulfone, 1-chloromethylsulfonyl benzene, chloromethyl sulphonyl benzene
C1=CC=C(C=C1)S(=O)(=O)CCl
190.641
190.64
Crystalline Powder
7205-98-3
White-Yellow
C7H7ClO2S
5 g
NXAIQSVCXQZNRY-UHFFFAOYSA-N
chloromethylsulfonylbenzene
81625
≥98.0% (GC)

Chemical Identifiers

7205-98-3
190.641
NXAIQSVCXQZNRY-UHFFFAOYSA-N
81625
C1=CC=C(C=C1)S(=O)(=O)CCl
C7H7ClO2S
MFCD00007551
chloromethyl phenyl sulfone, chloromethyl sulfonyl benzene, phenyl chloromethyl sulfone, chloromethylphenylsulfone, benzene, chloromethyl sulfonyl, chloromethylphenyl sulfone, 1-chloromethylsulfonyl benzene, chloromethyl sulphonyl benzene
chloromethylsulfonylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions