missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Phenyl Sulfone 98.0+%, TCI America™

Catalog No. p-7125066
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15715G 5 g
C157125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15715G Supplier TCI America Supplier No. C15715G
Only null left

Chemical Identifiers

CAS 7205-98-3
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.641
MDL Number MFCD00007551
InChI Key NXAIQSVCXQZNRY-UHFFFAOYSA-N
Synonym chloromethyl phenyl sulfone, chloromethyl sulfonyl benzene, phenyl chloromethyl sulfone, chloromethylphenylsulfone, benzene, chloromethyl sulfonyl, chloromethylphenyl sulfone, 1-chloromethylsulfonyl benzene, chloromethyl sulphonyl benzene
PubChem CID 81625
IUPAC Name chloromethylsulfonylbenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)CCl

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 130°C
Quantity 5 g
Formula Weight 190.64
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Chloromethyl Phenyl Sulfone