missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloromethyl Methyl Sulfide 97.0+%, TCI America™
Supplier: TCI America C060825G
Specifications
| Chloromethyl Methyl Sulfide | |
| Yellow | |
| C2H5ClS | |
| 25 g | |
| chloromethyl methyl sulfide, chlorodimethyl sulfide, methylthiomethyl chloride, chloro methylthio methane, methane, chloro methylthio, chlorodimethyl thioether, chloromethyl methyl thioether, monochlorodimethyl sulfide, chloro methylsulfanyl methane, clorodimetilsolfuro | |
| CSCCl | |
| 96.57 | |
| 96.57 | |
| Liquid |
| 2373-51-5 | |
| 106°C | |
| MFCD00000923 | |
| 1993 | |
| JWMLCCRPDOIBAV-UHFFFAOYSA-N | |
| chloro(methylsulfanyl)methane | |
| 16916 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 2373-51-5 | |
| 96.57 | |
| JWMLCCRPDOIBAV-UHFFFAOYSA-N | |
| 16916 | |
| CSCCl |
| C2H5ClS | |
| MFCD00000923 | |
| chloromethyl methyl sulfide, chlorodimethyl sulfide, methylthiomethyl chloride, chloro methylthio methane, methane, chloro methylthio, chlorodimethyl thioether, chloromethyl methyl thioether, monochlorodimethyl sulfide, chloro methylsulfanyl methane, clorodimetilsolfuro | |
| chloro(methylsulfanyl)methane |
Safety and Handling
RTECSNumber : WQ3700000
TSCA : No
Recommended Storage : Refrigerator