missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Methyl Sulfide 97.0+%, TCI America™

Catalog No. p-7124523
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C060825G 25 g
C0608100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C060825G Supplier TCI America Supplier No. C060825G
Only null left

Chemical Identifiers

CAS 2373-51-5
Molecular Formula C2H5ClS
Molecular Weight (g/mol) 96.57
MDL Number MFCD00000923
InChI Key JWMLCCRPDOIBAV-UHFFFAOYSA-N
Synonym chloromethyl methyl sulfide, chlorodimethyl sulfide, methylthiomethyl chloride, chloro methylthio methane, methane, chloro methylthio, chlorodimethyl thioether, chloromethyl methyl thioether, monochlorodimethyl sulfide, chloro methylsulfanyl methane, clorodimetilsolfuro
PubChem CID 16916
IUPAC Name chloro(methylsulfanyl)methane
SMILES CSCCl

Specifications

Color Yellow
Boiling Point 106°C
Quantity 25 g
UN Number 1993
Formula Weight 96.57
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloromethyl Methyl Sulfide
RTECSNumber WQ3700000
TSCA No
Recommended Storage Refrigerator