missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetic acid, 99%, flakes

CAS: 79-11-8 | C2H3ClO2 | 94.5 g/mol

Supplier:  Thermo Scientific Chemicals 108510010

 View more versions of this product

Catalog No. AC108510010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chloroacetic acid
79-11-8
61.0°C to 63.0°C
1.5800g/mL
126°C
99%
C2H3ClO2
MFCD00002683
02, 194
15, 2112
Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, chloroform, ether
C(C(=O)O)Cl
94.5
1.29 Pas (100°C)
94.5
99%
99%
250°C
White
189.0°C
Authentic
Plastic bottle
ClCH2CO2H
1 kg
1.58
chloroacetic acid, monochloroacetic acid, chloracetic acid, acetic acid, chloro, acide chloracetique, chloroethanoic acid, monochloroethanoic acid, monochloracetic acid, monochloorazijnzuur, monochloressigsaeure
FOCAUTSVDIKZOP-UHFFFAOYSA-N
2-chloroacetic acid
300
CHEBI:27869
0.75 mmHg at 20°C
Crystalline Flakes

Chemical Identifiers

79-11-8
94.5
FOCAUTSVDIKZOP-UHFFFAOYSA-N
300
2-chloroacetic acid
C2H3ClO2
MFCD00002683
chloroacetic acid, monochloroacetic acid, chloracetic acid, acetic acid, chloro, acide chloracetique, chloroethanoic acid, monochloroethanoic acid, monochloracetic acid, monochloorazijnzuur, monochloressigsaeure
CHEBI:27869
C(C(=O)O)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rins

GHS Signal Word: Danger

EINECSNumber : 201-178-4

RTECSNumber : AF8575000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only