missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroacetic acid, 99%, flakes
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Decomposition Information | 250°C |
| Melting Point | 61.0°C to 63.0°C |
| Color | White |
| Density | 1.5800g/mL |
| Boiling Point | 189.0°C |
| Flash Point | 126°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Plastic bottle |
| Linear Formula | ClCH2CO2H |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. May cause respiratory irritation. Toxic in contact with skin. Causes severe skin burns and eye damage. Fatal if inhaled. Very toxic to aquatic life. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF SWALLOWED: rins |
| EINECSNumber | 201-178-4 |
| RTECSNumber | AF8575000 |
| TSCA | TSCA |
RUO – Research Use Only