missing translation for 'onlineSavingsMsg'
Learn More

Chloro(triphenylphosphine)gold(I), Premion™, 99.99% (metals basis), Au 39.3% min

CAS: 14243-64-2 | C18H15AuClP | 494.71 g/mol

Supplier:  Thermo Scientific Chemicals 04100703

 View more versions of this product

Catalog No. AA4100703

Only null left
Explore available promotions

Description

Description

Chloro(triphenylphosphine)gold(I) is a common reagent in gold chemistry. It catalyzes certain rearrangement reactions in organic synthesis. It is also used to prepare 1-methylthyminato-N3-triphenylphosphinegold(I) by reacting with 1-methylthymine. It acts as a precursor for the synthesis of gold(I) and gold(III) organometallic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chloro(triphenylphosphine)gold(I)
Yellow
Odorless
MFCD00009588
chloro triphenylphosphine gold i, chloro triphenylphosphine gold, chlorotriphenylphosphine gold i, triphenylphosphinegold i chloride, aucl pph3, pubchem16017, ph3p aucl, chlorogold; triphenylphosphane, chlorogold; triphenylphosphine, chloranylgold; triphenylphosphane
IFPWCRBNZXUWGC-UHFFFAOYSA-M
chlorogold;triphenylphosphane
10874691
Powder
14243-64-2
99.99% (Metal basis, Au 39.3% min.)
C18H15AuClP
1 g
Soluble in methylene chloride,acetonitrile,benzene and acetone. Insoluble in water and ethanol.
Cl[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
494.71
494.71

Chemical Identifiers

14243-64-2
494.71
IFPWCRBNZXUWGC-UHFFFAOYSA-M
10874691
Cl[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
C18H15AuClP
MFCD00009588
chloro triphenylphosphine gold i, chloro triphenylphosphine gold, chlorotriphenylphosphine gold i, triphenylphosphinegold i chloride, aucl pph3, pubchem16017, ph3p aucl, chlorogold; triphenylphosphane, chlorogold; triphenylphosphine, chloranylgold; triphenylphosphane
chlorogold;triphenylphosphane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 238-117-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only