missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloro(methyl)(phenyl)(vinyl)silane 98.0+%, TCI America™
$180.81 - $659.87
Chemical Identifiers
| CAS | 17306-05-7 |
|---|---|
| Molecular Formula | C9H11ClSi |
| Molecular Weight (g/mol) | 182.722 |
| MDL Number | MFCD00053208 |
| InChI Key | GSXJAPJSIVGONK-UHFFFAOYSA-N |
| PubChem CID | 87042 |
| IUPAC Name | chloro-ethenyl-methyl-phenylsilane |
| SMILES | C[Si](C=C)(C1=CC=CC=C1)Cl |
Chemical Identifiers
| 17306-05-7 | |
| 182.722 | |
| GSXJAPJSIVGONK-UHFFFAOYSA-N | |
| chloro-ethenyl-methyl-phenylsilane |
| C9H11ClSi | |
| MFCD00053208 | |
| 87042 | |
| C[Si](C=C)(C1=CC=CC=C1)Cl |
Specifications
| 17306-05-7 | |
| 80°C | |
| MFCD00053208 | |
| 2987 | |
| C[Si](C=C)(C1=CC=CC=C1)Cl | |
| 182.722 | |
| 182.72 | |
| Liquid |
| Yellow | |
| C9H11ClSi | |
| 5 g | |
| GSXJAPJSIVGONK-UHFFFAOYSA-N | |
| chloro-ethenyl-methyl-phenylsilane | |
| 87042 | |
| ≥98.0% (GC) | |
| Chloro(methyl)(phenyl)(vinyl)silane |
Safety and Handling
EINECSNumber : (3)-1165
TSCA : Yes