missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloro(methyl)(phenyl)(vinyl)silane 98.0+%, TCI America™
Supplier: TCI America C343425G
Specifications
| Chloro(methyl)(phenyl)(vinyl)silane | |
| Yellow | |
| C9H11ClSi | |
| 25 g | |
| GSXJAPJSIVGONK-UHFFFAOYSA-N | |
| chloro-ethenyl-methyl-phenylsilane | |
| 87042 | |
| ≥98.0% (GC) |
| 17306-05-7 | |
| 80°C | |
| MFCD00053208 | |
| 2987 | |
| C[Si](C=C)(C1=CC=CC=C1)Cl | |
| 182.722 | |
| 182.72 | |
| Liquid |
Chemical Identifiers
| 17306-05-7 | |
| 182.722 | |
| GSXJAPJSIVGONK-UHFFFAOYSA-N | |
| chloro-ethenyl-methyl-phenylsilane |
| C9H11ClSi | |
| MFCD00053208 | |
| 87042 | |
| C[Si](C=C)(C1=CC=CC=C1)Cl |
Safety and Handling
EINECSNumber : (3)-1165
TSCA : Yes