accessibility menu, dialog, popup

UserName

H3CSiClH2C

CAS RN 17306-05-7

IUPAC Name: chloro(ethenyl)methyl(phenyl)silane
Synonyms: chloro(ethenyl)methyl(phenyl)silane
Molecular Weight (g/mol): 182.72
Molecular Formula: C9H11ClSi
InChi Key: GSXJAPJSIVGONK-UHFFFAOYNA-N
SMILES: C[Si](Cl)(C=C)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results