missing translation for 'onlineSavingsMsg'
Learn More

Chlorine Water, Saturated, Approximately 0.3% (w/v), Ricca Chemical

Catalog No. p-3251490
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Safety-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
198016 500 mL Safety-coated Glass Bottle
198032 1 L Safety-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 198016 Supplier Ricca Chemical Company Supplier No. RI198016
Only null left

Chemical Identifiers

CAS 7782-50-5 , 7732-18-5
Molecular Formula Cl2
Molecular Weight (g/mol) 70.9
InChI Key KZBUYRJDOAKODT-UHFFFAOYSA-N
Synonym chlorine, bertholite, chloor, cloro, chlorine gas, chlorine mol., chlore french, cloro italian, chloor dutch, chlor german
PubChem CID 24526
ChEBI CHEBI:29310
IUPAC Name molecular chlorine
SMILES ClCl

Specifications

Color Green-Yellow
CAS Min % 98.0
CAS Max % 100.0
Grade Laboratory
Packaging Safety-coated Glass Bottle
Solvent or Matrix Saturated Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes approximately 0.3% (w/v)
Chemical Name or Material Chlorine Water