missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloride Standard for Ion Chromatography, SPEX CertiPrep™
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Cl&superscript_minus; | 0.1M |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Packaging | Triple-leached LDPE bottle |
| Certificate of Analysis | With Certificate of Analysis |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| SMILES | [NH4+].[Cl-] |
| Solvent or Matrix | Water |
| Quantity | 125 mL |
| Concentration or Composition (by Analyte or Components) | 0.1M |
| Chemical Name or Material | Chloride Standard |
Safety and Handling
| DOTInformation | Non-Hazardous |
| Recommended Storage | Ambient |
Product intended for laboratory use only