missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloride Standard, 1000 ppm Cl-, Fisherbrand
Supplier: Ricca Chemical Company ACCU0819500A
Specifications
Chloride Standard | |
Natural Poly Bottle | |
Odorless | |
ClNa | |
Colorless | |
100°C | |
Calibration Standard | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
500 mL | |
CHEBI:26710 |
1000ppm (Cl) | |
7647-14-5,7732-18-5 | |
Liquid | |
MFCD00003477 | |
0.0°C | |
Miscible with water | |
Single-component Standard | |
[Na+].[Cl-] | |
58.44 | |
5234 | |
Laboratory |
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
CHEBI:26710 | |
[Na+].[Cl-] |
ClNa | |
MFCD00003477 | |
5234 | |
sodium chloride |