missing translation for 'onlineSavingsMsg'
Learn More

Chloride Standard, 10 ppm Cl-, Ricca Chemical

Supplier:  Ricca Chemical Company R19421001A

 View more versions of this product

Catalog No. R19421001A


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

CHLORIDE STANDARD
ClH4N
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
N.Cl
53.49
CHEBI:31206
Natural Poly Bottle
1 L
12125-02-9
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
amine hydrochloride
25517
Laboratory
6.7 to 7.3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions