missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloride Standard, 10 ppm Cl-, Ricca Chemical
$100.19 - $235.97
Specifications
| Chemical Name or Material | CHLORIDE STANDARD |
|---|---|
| CAS | 12125-02-9 |
| Molecular Formula | ClH4N |
| MDL Number | MFCD00011420 |
| Synonym | ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
19421-16
|
Ricca Chemical Company
1942116 |
500 mL |
Each for $174.22
|
|
|||||
|
R19421001A
|
Ricca Chemical Company
R19421001A |
1 L |
Each for $235.97
|
|
|||||
|
R19421004A
|
Ricca Chemical Company
R19421004A |
4 L |
Each for $100.19
|
|
|||||
Specifications
| CHLORIDE STANDARD | |
| ClH4N | |
| ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
| N.Cl | |
| 53.49 | |
| CHEBI:31206 | |
| Natural Poly Bottle |
| 12125-02-9 | |
| MFCD00011420 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| amine hydrochloride | |
| 25517 | |
| Laboratory | |
| 6.7 to 7.3 |