missing translation for 'onlineSavingsMsg'
Learn More

Chloramine-T, 1% (w/v), Ricca Chemical

Catalog No. 192616
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
192616 500 mL Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 192616 Supplier Ricca Chemical Company Supplier No. 192616
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7080-50-4
Molecular Formula C7H7ClNNaO2S
Molecular Weight (g/mol) 227.638
InChI Key VDQQXEISLMTGAB-UHFFFAOYSA-N
PubChem CID 3641960
ChEBI CHEBI:53767
IUPAC Name sodium;chloro-(4-methylphenyl)sulfonylazanide
SMILES CC1=CC=C(C=C1)S(=O)(=O)[N-]Cl.[Na+]

Specifications

Type Chloramine T
Physical Form Liquid
CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Packaging Amber Poly Bottle
Color Colorless to Yellow
Concentration 1% (w/v)
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Chloramine-T