missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloramine B Hydrate 80.0+%, TCI America™
$80.16 - $80.16
Chemical Identifiers
| CAS | 127-52-6 |
|---|---|
| Molecular Formula | C6H5ClNNaO2S |
| Molecular Weight (g/mol) | 213.61 |
| MDL Number | MFCD01459908 |
| InChI Key | FTDKYSGFGRDGGE-UHFFFAOYSA-M |
| Synonym | Benzenesulfonechloramide Sodium Salt |
| PubChem CID | 22141507 |
| IUPAC Name | sodium (chloroimino)(phenyl)-λâ¶-sulfanoylolate |
| SMILES | [Na+].[O-]S(=O)(=NCl)C1=CC=CC=C1 |
Chemical Identifiers
| 127-52-6 | |
| 213.61 | |
| FTDKYSGFGRDGGE-UHFFFAOYSA-M | |
| 22141507 | |
| [Na+].[O-]S(=O)(=NCl)C1=CC=CC=C1 |
| C6H5ClNNaO2S | |
| MFCD01459908 | |
| Benzenesulfonechloramide Sodium Salt | |
| sodium (chloroimino)(phenyl)-λâ¶-sulfanoylolate |
Specifications
| 127-52-6 | |
| C6H5ClNNaO2S | |
| 25 g | |
| Benzenesulfonechloramide Sodium Salt | |
| [Na+].[O-]S(=O)(=NCl)C1=CC=CC=C1 | |
| 213.61 | |
| 213.61 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD01459908 | |
| 1479 | |
| FTDKYSGFGRDGGE-UHFFFAOYSA-M | |
| sodium (chloroimino)(phenyl)-λâ¶-sulfanoylolate | |
| 22141507 | |
| ≥80.0% (T) | |
| Chloramine B Hydrate |
Safety and Handling
RTECSNumber : DA9300000
TSCA : Yes