accessibility menu, dialog, popup

UserName

NaOSONCl

CAS RN 127-52-6

IUPAC Name: sodium (chloroimino)(phenyl)-λ⁶-sulfanoylolate
Synonyms: sodium (chloroimino)(phenyl)-λ⁶-sulfanoylolate
Molecular Weight (g/mol): 213.61
Molecular Formula: C6H5ClNNaO2S
InChi Key: FTDKYSGFGRDGGE-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=NCl)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results