missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CHKtide

Catalog No. PV5368
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5368 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV5368 Supplier Thermo Scientific™ Supplier No. PV5368
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A peptide substrate based upon CDC25C that is phosphorylated by CHEK1 and other kinases.

A peptide substrate based upon CDC25C that is phosphorylated by CHEK1 and other kinases.

For use with the Adapta™ Universal Kinase Assay (Cat. No. PV5099).

Specifications

Amino Acid Sequence KKKVSRSGLYRSPSMPENLNRPR
For Use With (Application) Kinase Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Pharma and Biopharma, Target and Lead Identification and Validation
Formulation CHKtide peptide substrate in in 50mM HEPES pH 7.5, 0.01% Brij-35, 10mM MgCl2 and 1mM EGTA.
Molecular Weight (g/mol) 2701.2 Da
Content And Storage 2mM CHKtide peptide substrate in 50 mM HEPES pH 7.5, 0.01% Brij-35, 10 mM MgCl2, and 1 mM EGTA

Store at -20°C.
Detection Method Fluorescence
Form Frozen
Quantity 1 mL
Storage Requirements Store at -20°C.
Substrate peptide based upon CDC25C
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Structural Form KKKVSRSGLYRSPSMPENLNRPR
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.