missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CHES, 99%

Catalog No. AAA1804714
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1804714 25 g
AAA1804722 100 g
AAA1804736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1804714 Supplier Thermo Scientific Chemicals Supplier No. A1804714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol

CHES is zwitterionic buffer useful in pH range 8.6 to 10.0.CHES is widely utilized as a buffer which is used for investigations on pH-dependent processes in enzymology. It has been shown to have an unusually high affinity for the iodoacetate binding site of liver alcohol dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
CHES is zwitterionic buffer useful in pH range 8.6 to 10.0.CHES is widely utilized as a buffer which is used for investigations on pH-dependent processes in enzymology. It has been shown to have an unusually high affinity for the iodoacetate binding site of liver alcohol dehydrogenase.

Solubility
Soluble in water.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 103-47-9
Molecular Formula C8H17NO3S
Molecular Weight (g/mol) 207.288
MDL Number MFCD00003835
InChI Key MKWKNSIESPFAQN-UHFFFAOYSA-N
Synonym 2-(Cyclohexylamino)ethanesulfonic acid
PubChem CID 66898
IUPAC Name 2-(cyclohexylamino)ethanesulfonic acid
SMILES C1CCC(CC1)NCCS(=O)(=O)O

Specifications

Melting Point >300°C
Quantity 25 g
Assay Percent Range 0.99
Beilstein 2967601
Merck Index 14,2055
Solubility Information Soluble in water.
Formula Weight 207.29
Percent Purity 99%
Chemical Name or Material CHES
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
EINECSNumber 203-115-6
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only