missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CHES, 98+%, for biochemistry

Catalog No. AC208180250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC208180250 25 g Plastic bottle
AC208181000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC208180250 Supplier Thermo Scientific Chemicals Supplier No. 208180250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 103-47-9
Molecular Formula C8H17NO3S
Molecular Weight (g/mol) 207.288
MDL Number MFCD00003835
InChI Key MKWKNSIESPFAQN-UHFFFAOYSA-N
Synonym 2-(Cyclohexylamino)ethanesulfonic acid
PubChem CID 66898
IUPAC Name 2-(cyclohexylamino)ethanesulfonic acid
SMILES C1CCC(CC1)NCCS(=O)(=O)O

Specifications

Absorbance 0.05 max. at 260nm
Absorbance Conditions (0.1M aq. soln.)
Melting Point 300.0°C
Color White
pH 5 to 6 (1% aq. soln.)
Physical Form Fine Crystalline Powder
Quantity 25 g
Assay Percent Range 98+%
Infrared Spectrum Authentic
Linear Formula C6H11NHCH2CH2SO3H
Packaging Plastic bottle
Merck Index 15, 2055
Solubility Information Solubility in water: 103.6g/L (20°C).
Formula Weight 207.29
Percent Purity ≥98%
Grade Biochemical
Chemical Name or Material CHES, 98+%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-115-6
TSCA TSCA

RUO – Research Use Only