Learn More
Thermo Scientific Chemicals CHES, 0.5M buffer soln, pH 9.0
CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol
$174.25 - $282.45
Chemical Identifiers
| Molecular Formula | C8H17NO3S |
|---|---|
| Molecular Weight (g/mol) | 207.288 |
| MDL Number | MFCD00003835 |
| InChI Key | MKWKNSIESPFAQN-UHFFFAOYSA-N |
| PubChem CID | 66898 |
| IUPAC Name | 2-(cyclohexylamino)ethanesulfonic acid |
| SMILES | C1CCC(CC1)NCCS(=O)(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ63670AE
|
Thermo Scientific Chemicals
J63670AE |
100 mL |
Each for $174.25
|
|
|||||
|
AAJ63670AK
|
Thermo Scientific Chemicals
J63670AK |
250 mL |
Each for $282.45
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C8H17NO3S | |
| MFCD00003835 | |
| 66898 | |
| C1CCC(CC1)NCCS(=O)(=O)O |
| 207.288 | |
| MKWKNSIESPFAQN-UHFFFAOYSA-N | |
| 2-(cyclohexylamino)ethanesulfonic acid |
Specifications
| 0.5 M | |
| Liquid | |
| C8H17NO3S | |
| MKWKNSIESPFAQN-UHFFFAOYSA-N | |
| 2-(cyclohexylamino)ethanesulfonic acid | |
| 66898 |
| 9.0 | |
| 100 mL | |
| MFCD00003835 | |
| C1CCC(CC1)NCCS(=O)(=O)O | |
| 207.288 | |
| CHES buffer soln. |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only