missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coverall with Serged Seams, Attached Hood and Boot, Elastic Wrist
These durable and disposable coveralls with serged seams and elastic back protect against chemical splashes and toxic particles.
$139.28 - $191.25
Specifications
| Product Type | Chemical Protective Clothing |
|---|---|
| Gender | Unisex |
| Material | Laminated Non-woven Fabric |
| Color | Yellow |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-184-112
|
International Enviroguard™
7019YSM |
Medium |
Case of 12 Each for $139.28
|
|
|||||
|
17-184-113
|
International Enviroguard™
7019YSL |
Large |
Case of 12 Each for $142.44
|
|
|||||
|
17-184-111
|
International Enviroguard™
7019YSXL |
X-Large |
Case of 12 Each for $148.84
|
|
|||||
|
17-184-117
|
International Enviroguard™
7019YS2XL |
2X-Large |
Case of 12 Each for $155.37
|
|
|||||
|
17-184-116
|
International Enviroguard™
7019YS3XL |
3X-Large |
Case of 12 Each for $160.03
|
|
|||||
|
17-184-115
|
International Enviroguard™
7019YS4XL |
4X-Large |
Case of 12 Each for $171.08
|
|
|||||
|
17-184-114
|
International Enviroguard™
7019YS5XL |
5X-Large |
Case of 12 Each for $191.25
|
|
|||||
Description
- Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
- A storm flap over the zipper with pressure-sensitive tape helps prevent hazardous liquids from seeping through exposed zipper seams
- For added comfort, seamless shoulders help prevent tears and allow you to move more freely as you work
- With an attached hood and boots, you're protected from head to toe
- Wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
- Elastic back stretches with user for more comfort and better tear resistance
- Elastic wrists offer a secure fit to keep sleeves in place to decrease the likelihood of skin exposure
- Possible applications include manufacturing, emergency response, environmental cleanup, and more
Specifications
| Chemical Protective Clothing | |
| Laminated Non-woven Fabric | |
| Single-use | |
| Attached Boots | |
| Hooded | |
| Serged | |
| Acid, Caustics, Chemicals, Isopropyl Alcohol, Particle | |
| None |
| Unisex | |
| Yellow | |
| Elastic at Back | |
| Elastic | |
| 2-way Front Zipper with Storm Flap | |
| Non-sterile | |
| Yes |