missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coverall with Serged Seams, Attached Hood and Boot, Elastic Wrist
These durable and disposable coveralls with serged seams and elastic back protect against chemical splashes and toxic particles.
Supplier: International Enviroguard™ 7019YSXL
Description
- Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
- A storm flap over the zipper with pressure-sensitive tape helps prevent hazardous liquids from seeping through exposed zipper seams
- For added comfort, seamless shoulders help prevent tears and allow you to move more freely as you work
- With an attached hood and boots, you're protected from head to toe
- Wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
- Elastic back stretches with user for more comfort and better tear resistance
- Elastic wrists offer a secure fit to keep sleeves in place to decrease the likelihood of skin exposure
- Possible applications include manufacturing, emergency response, environmental cleanup, and more
Specifications
| Chemical Protective Clothing | |
| Unisex | |
| Yellow | |
| Elastic at Back | |
| Elastic | |
| Attached, Elastic Face | |
| Serged | |
| Acid, Caustics, Chemicals, Isopropyl Alcohol, Particle | |
| None |
| X-Large | |
| Laminated Non-woven Fabric | |
| Single-use | |
| Attached Boots | |
| Hooded | |
| 2-way Front Zipper with Storm Flap | |
| Non-sterile | |
| Yes |