missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coverall with Serged Seams, Attached Hood and Boot, Elastic Wrist
Description
- Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
- A storm flap over the zipper with pressure-sensitive tape helps prevent hazardous liquids from seeping through exposed zipper seams
- For added comfort, seamless shoulders help prevent tears and allow you to move more freely as you work
- With an attached hood and boots, you're protected from head to toe
- Wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
- Elastic back stretches with user for more comfort and better tear resistance
- Elastic wrists offer a secure fit to keep sleeves in place to decrease the likelihood of skin exposure
- Possible applications include manufacturing, emergency response, environmental cleanup, and more
Specifications
Specifications
| Product Type | Chemical Protective Clothing |
| Size | Large |
| Gender | Unisex |
| Material | Laminated Non-woven Fabric |
| Color | Yellow |
| Disposable | Single-use |
| Waist Style | Elastic at Back |
| Ankle Style | Attached Boots |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |