missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ QCM™ ECMatrix Cell Invasion Assays
For evaluation of invasive tumor cells
$897.00
Specifications
| Content And Storage | 2°C to 8°C |
|---|---|
| Description | QCM ECMatrix Cell Invasion Assay |
| Detection Method | Fluorescence |
| For Use With (Application) | Evaluation of invasive tumor cells |
Specifications
| 2°C to 8°C | |
| QCM ECMatrix Cell Invasion Assay | |
| Fluorescence | |
| Evaluation of invasive tumor cells |