missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ QCM™ ECMatrix Cell Invasion Assays

Catalog No. ECM554
Change view
Click to view available options
Format:
24-well
96-well
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
ECM554 24-well
ECM555 96-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ECM554 Supplier MilliporeSigma™ Supplier No. ECM554

For evaluation of invasive tumor cells

Specifications

Content And Storage 2°C to 8°C
Description QCM ECMatrix Cell Invasion Assay
Detection Method Fluorescence
For Use With (Application) Evaluation of invasive tumor cells
Format 24-well
Includes Sterile 24-well plate assembly, cell detachment solution, 4X cell lysis buffer, CyQuant GR Dye 1
Quantity 24 Assays