missing translation for 'onlineSavingsMsg'
Learn More

Chelerythrine chloride, 99+%

CAS: 3895-92-9 | C21H18ClNO4 | 383.83 g/mol

Supplier:  Thermo Scientific Chemicals J62906MA

 View more versions of this product

Catalog No. AAJ62906MA


Only null left
Explore available promotions

Description

Description

A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A natural benzophenanthridine alkaloid
  • Inhibits protein kinase C
  • Induces apoptosis in leukemia cells
Specifications

Specifications

Chelerythrine chloride
Yellow to Orange
MFCD00060717
14,2051
Soluble in DMSO at 2mg/ml
[Cl-].COC1=CC=C2C(C=[N+](C)C3=C2C=CC2=CC4=C(OCO4)C=C32)=C1OC
383.83
383.82
Solid
3895-92-9
C21H18ClNO4
10 mg
chelerythrine chloride, chelerythrine hydrochloride, chelerythrinechloride, unii-7ic98tz0pz, 1,2-dimethoxy-12-methyl-1,3 dioxolo 4',5':4,5 benzo 1,2-c phenanthridin-12-ium chloride, chelerythrine, chloride, 7ic98tz0pz, 1,2-dimethoxy-n-methyl-1,3-benzodioxolo 5,6c phenanthridinium, 1,2-dimethoxy-12-methyl 1,3 benzodioxolo 5,6-c phenanthridinium chloride
WEEFNMFMNMASJY-UHFFFAOYSA-M
1,2-dimethoxy-12-methyl-[1,3]benzodioxolo[5,6-c]phenanthridin-12-ium;chloride
72311
≥99%

Chemical Identifiers

3895-92-9
383.83
WEEFNMFMNMASJY-UHFFFAOYSA-M
72311
C21H18ClNO4
MFCD00060717
chelerythrine chloride, chelerythrine hydrochloride, chelerythrinechloride, unii-7ic98tz0pz, 1,2-dimethoxy-12-methyl-1,3 dioxolo 4',5':4,5 benzo 1,2-c phenanthridin-12-ium chloride, chelerythrine, chloride, 7ic98tz0pz, 1,2-dimethoxy-n-methyl-1,3-benzodioxolo 5,6c phenanthridinium, 1,2-dimethoxy-12-methyl 1,3 benzodioxolo 5,6-c phenanthridinium chloride
[Cl-].COC1=CC=C2C(C=[N+](C)C3=C2C=CC2=CC4=C(OCO4)C=C32)=C1OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319

EINECSNumber : 223-444-9

RTECSNumber : FL9200000

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only