Learn More
Chelerythrine chloride, 99+%
CAS: 3895-92-9 | C21H18ClNO4 | 383.83 g/mol
$264.79 - $854.41
Chemical Identifiers
| CAS | 3895-92-9 |
|---|---|
| Molecular Formula | C21H18ClNO4 |
| Molecular Weight (g/mol) | 383.83 |
| MDL Number | MFCD00060717 |
| InChI Key | WEEFNMFMNMASJY-UHFFFAOYSA-M |
| Synonym | chelerythrine chloride, chelerythrine hydrochloride, chelerythrinechloride, unii-7ic98tz0pz, 1,2-dimethoxy-12-methyl-1,3 dioxolo 4',5':4,5 benzo 1,2-c phenanthridin-12-ium chloride, chelerythrine, chloride, 7ic98tz0pz, 1,2-dimethoxy-n-methyl-1,3-benzodioxolo 5,6c phenanthridinium, 1,2-dimethoxy-12-methyl 1,3 benzodioxolo 5,6-c phenanthridinium chloride |
| PubChem CID | 72311 |
| SMILES | [Cl-].COC1=CC=C2C(C=[N+](C)C3=C2C=CC2=CC4=C(OCO4)C=C32)=C1OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ62906LB0
|
Thermo Scientific Chemicals
J62906LB0 |
5 mg |
Each for $264.79
|
|
|||||
|
AAJ62906MA
|
Thermo Scientific Chemicals
J62906MA |
10 mg |
Each for $407.78
|
|
|||||
|
AAJ62906MB
|
Thermo Scientific Chemicals
J62906MB |
25 mg |
Each for $854.41
|
|
|||||
Description
A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A natural benzophenanthridine alkaloid
- Inhibits protein kinase C
- Induces apoptosis in leukemia cells
Chemical Identifiers
| 3895-92-9 | |
| 383.83 | |
| WEEFNMFMNMASJY-UHFFFAOYSA-M | |
| 72311 |
| C21H18ClNO4 | |
| MFCD00060717 | |
| chelerythrine chloride, chelerythrine hydrochloride, chelerythrinechloride, unii-7ic98tz0pz, 1,2-dimethoxy-12-methyl-1,3 dioxolo 4',5':4,5 benzo 1,2-c phenanthridin-12-ium chloride, chelerythrine, chloride, 7ic98tz0pz, 1,2-dimethoxy-n-methyl-1,3-benzodioxolo 5,6c phenanthridinium, 1,2-dimethoxy-12-methyl 1,3 benzodioxolo 5,6-c phenanthridinium chloride | |
| [Cl-].COC1=CC=C2C(C=[N+](C)C3=C2C=CC2=CC4=C(OCO4)C=C32)=C1OC |
Specifications
| 3895-92-9 | |
| C21H18ClNO4 | |
| 5 mg | |
| chelerythrine chloride, chelerythrine hydrochloride, chelerythrinechloride, unii-7ic98tz0pz, 1,2-dimethoxy-12-methyl-1,3 dioxolo 4',5':4,5 benzo 1,2-c phenanthridin-12-ium chloride, chelerythrine, chloride, 7ic98tz0pz, 1,2-dimethoxy-n-methyl-1,3-benzodioxolo 5,6c phenanthridinium, 1,2-dimethoxy-12-methyl 1,3 benzodioxolo 5,6-c phenanthridinium chloride | |
| WEEFNMFMNMASJY-UHFFFAOYSA-M | |
| 1,2-dimethoxy-12-methyl-[1,3]benzodioxolo[5,6-c]phenanthridin-12-ium;chloride | |
| 72311 | |
| ≥99% | |
| Chelerythrine chloride |
| Yellow to Orange | |
| MFCD00060717 | |
| 14,2051 | |
| Soluble in DMSO at 2mg/ml | |
| [Cl-].COC1=CC=C2C(C=[N+](C)C3=C2C=CC2=CC4=C(OCO4)C=C32)=C1OC | |
| 383.83 | |
| 383.82 | |
| Solid |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
EINECSNumber : 223-444-9
RTECSNumber : FL9200000
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only