missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CGI 1746

Catalog No. 696510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
696510 10 mg
696550 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 696510 Supplier Tocris Bioscience™ Supplier No. 6965/10
Only null left

Potent, reversible BTK inhibitor

CGI 1746 is a potent, reversible inhibitor of Bruton's tyrosine kinase (Btk) (IC50 = 1.9 nM). CGI 1746 decreases cell survival in prostate cancer cell lines. In a cell model, CGI 1746 blocks the production of FcγR-induced cytokines IL-1β, TNFα and IL-6 (IC50 values are 36 nM, 47 nM and 353 nM, respectively).

Specifications

Chemical Name or Material N-[3-[4,5-Dihydro-4-methyl-6-[[4-(4-morpholinylcarbonyl)phenyl]amino]-5-oxo-2-pyrazinyl]-2-methylphenyl]-4-(1,1-dimethylethyl)benzamide
CAS 910232-84-7
Quantity 10 mg
Target Brutons Tyrosine Kinase (BTK) Inhibitors
Molecular Formula C34H37N5O4
Purity 0.97