missing translation for 'onlineSavingsMsg'
Learn More

Cetyltrimethylammonium Bromide, 98-101%, Spectrum™ Chemical

CE122, 57-09-0, C19H42BrN

Supplier:  Spectrum Chemical Mfg Cor CE12225KG

 View more versions of this product

Catalog No. 18601568


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Cetyltrimethylammonium Bromide is also referred to as CTAB and is a cationic surfactant that is used as a buffer solution during the extraction of DNA, in hair conditioning products, as well as, in the synthesis of gold nanoparticles.
Specifications

Specifications

57-09-0
Poly Pail
MFCD00011772
LZZYPRNAOMGNLH-UHFFFAOYSA-M
hexadecyltrimethylazanium bromide
98-101%
100%
C19H42BrN
2.5 kg
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
364.46
Reagent

Chemical Identifiers

57-09-0
364.46
LZZYPRNAOMGNLH-UHFFFAOYSA-M
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
C19H42BrN
MFCD00011772
hexadecyltrimethylazanium bromide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions