missing translation for 'onlineSavingsMsg'
Learn More

Cetyltrimethylammonium Bromide, 98-101%, Spectrum™ Chemical

Catalog No. p-7157259
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601566 125 g Amber Glass Bottle
18601567 500 g Amber Glass Bottle
18601568 2.5 kg Poly Pail
18601569 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601566 Supplier Spectrum Chemical Mfg Cor Supplier No. CE122125GM
Only null left

CE122, 57-09-0, C19H42BrN

Spectrum™ Chemical Cetyltrimethylammonium Bromide is also referred to as CTAB and is a cationic surfactant that is used as a buffer solution during the extraction of DNA, in hair conditioning products, as well as, in the synthesis of gold nanoparticles.

Chemical Identifiers

CAS 57-09-0
Molecular Formula C19H42BrN
Molecular Weight (g/mol) 364.46
MDL Number MFCD00011772
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
IUPAC Name hexadecyltrimethylazanium bromide
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 125 g
Percent Purity 98-101%
Grade Reagent