missing translation for 'onlineSavingsMsg'
Learn More

Cesium sulfate, 99%

CAS: 10294-54-9 | Cs2O4S | 361.87 g/mol

Supplier:  Thermo Scientific Chemicals A1676736

 View more versions of this product

Catalog No. AAA1676736


Only null left
Explore available promotions

Description

Description

Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.

Solubility
Soluble in water. (1820 g/L) at 20°C

Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water, reducing agents and oxidizing agents.
Specifications

Specifications

Cesium sulfate
10294-54-9
Cs2O4S
14,2016
Soluble in water. (1820g/L) at 20°C
[Cs+].[Cs+].[O-]S([O-])(=O)=O
361.87
361.87
Odorless
4.24
1,010°C
500 g
MFCD00010959
cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
FLJPGEWQYJVDPF-UHFFFAOYSA-L
dicesium;sulfate
25137
99%
Hygroscopic

Chemical Identifiers

10294-54-9
361.87
FLJPGEWQYJVDPF-UHFFFAOYSA-L
25137
[Cs+].[Cs+].[O-]S([O-])(=O)=O
Cs2O4S
MFCD00010959
cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
dicesium;sulfate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H361f-H373

EINECSNumber : 233-662-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only