missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium sulfate, 99%
Description
Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.
Solubility
Soluble in water. (1820 g/L) at 20°C
Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water, reducing agents and oxidizing agents.
Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.
Solubility
Soluble in water. (1820 g/L) at 20°C
Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water, reducing agents and oxidizing agents.
Specifications
Specifications
| Melting Point | 1,010°C |
| Quantity | 25 g |
| Merck Index | 14,2016 |
| Solubility Information | Soluble in water. (1820g/L) at 20°C |
| Formula Weight | 361.87 |
| Percent Purity | 99% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 4.24 |
| Chemical Name or Material | Cesium sulfate |
Safety and Handling
| Hazard Category | H302-H361f-H373 |
| Precautionary Statement | P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c |
| EINECSNumber | 233-662-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only