accessibility menu, dialog, popup

UserName

CsCsOSOOO

CAS RN 10294-54-9

IUPAC Name: dicaesium(1+) sulfate
Synonyms: cesium sulfate|caesium sulfate
Molecular Weight (g/mol): 361.87
Molecular Formula: Cs2O4S
InChi Key: FLJPGEWQYJVDPF-UHFFFAOYSA-L
SMILES: [Cs+].[Cs+].[O-]S([O-])(=O)=O
Quantity 
  • (2)
  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)
  • (4)
  • (6)
Percent Purity 
  • (4)
  • (3)
  • (4)
  • (2)
Grade 
  • (2)
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (13)
  • (2)
  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (6)
  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (4)
  • (4)