missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Sulfate, 99.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor C122510KGBL
Specifications
| 10294-54-9 | |
| 10 kg | |
| MFCD00010959 | |
| [Cs+].[Cs+].[O-]S([O-])(=O)=O | |
| 361.87 | |
| Reagent |
| 1 | |
| Cs2O4S | |
| FLJPGEWQYJVDPF-UHFFFAOYSA-L | |
| dicaesium(1+) sulfate | |
| 99.5% | |
| Poly Pail |
Chemical Identifiers
| 10294-54-9 | |
| 361.87 | |
| FLJPGEWQYJVDPF-UHFFFAOYSA-L | |
| [Cs+].[Cs+].[O-]S([O-])(=O)=O |
| Cs2O4S | |
| MFCD00010959 | |
| dicaesium(1+) sulfate |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.