missing translation for 'onlineSavingsMsg'
Learn More

Cesium Sulfate, 99.5%, Spectrum™ Chemical

Catalog No. p-7165316
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
10 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609649 5 g Amber Glass Bottle
18609648 25 g Amber Glass Bottle
18609647 100 g Amber Glass Bottle
18609646 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609649 Supplier Spectrum Chemical Mfg Cor Supplier No. C12255GM
Only null left

Chemical Identifiers

CAS 10294-54-9
Molecular Formula Cs2O4S
Molecular Weight (g/mol) 361.87
MDL Number MFCD00010959
InChI Key FLJPGEWQYJVDPF-UHFFFAOYSA-L
IUPAC Name dicaesium(1+) sulfate
SMILES [Cs+].[Cs+].[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 5 g
Percent Purity 99.5%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.