Learn More
Fisherbrand™ Certified Low-Particulate Clear French Squares
GREENER_CHOICE
With PTFE-lined caps
$934.10 - $939.55
Specifications
Color Clear
Material Glass
Closure Material PTFE
Closure Type Screw Cap
View More Specs
Products 2
Catalog Number Capacity (Metric) Price Quantity & Availability  
Catalog Number Capacity (Metric) Price Quantity & Availability  
02911890
GREENER_CHOICE
View Documents Promotion Details
125 mL
Case of 24 Each for $934.10
Only null left
02911893
GREENER_CHOICE
View Documents Promotion Details
250 mL
Case of 24 Each for $939.55
Only null left
Description
Description
  • Containers and caps are cleaned to five particles or less per milliliter at 0.5 microns with USP Purified Water or Water for Injection in a Class 100/10 cleanroom
Order Info
Custom processing, packaging and certification are available by special request
Certifications
Certificate of Analysis is included with each case.
Specifications
Specifications
Clear
Glass
PTFE
Screw Cap