missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceric Sulfate, 0.100 Normal (N/10) In Dilute Sulfuric Acid, Reagents
Ceric Sulfate, 0.100 Normal (N/10) In Dilute Sulfuric Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
Supplier: Labchem Inc C7172200500A
Specifications
| 100°C | |
| Orange | |
| <1 | |
| 500 mL | |
| Cerium(IV) Sulfate | |
| O | |
| 18.02 | |
| Poly Bottle |
| Ceric Sulfate, 0.100 Normal (N/10) In Dilute Sulfuric Acid | |
| 7732-18-5,7664-93-9,7664-93-9 | |
| Liquid | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent | |
| 1.085 |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Cerium(IV) Sulfate | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |