missing translation for 'onlineSavingsMsg'
Learn More

Ceric Sulfate, 0.100 Normal (N/10) In Dilute Sulfuric Acid, Reagents

Catalog No. p-200021756
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336525 10 L
01336524 1 L
01336523 20 L
01336522 4 L
01336521 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01336525 Supplier Labchem Inc Supplier No. C717220010F
Only null left

Ceric Sulfate, 0.100 Normal (N/10) In Dilute Sulfuric Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Cerium(IV) Sulfate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Orange
pH <1
Physical Form Liquid
Quantity 10 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.085
Chemical Name or Material Ceric Sulfate, 0.100 Normal (N/10) In Dilute Sulfuric Acid