missing translation for 'onlineSavingsMsg'
Learn More

Cephalexin Monohydrate 98.0+%, TCI America™

Supplier:  TCI America C224825G

 View more versions of this product

Catalog No. C224825G


Only null left

Description

Description

Specifications

Specifications

Cephalexin Monohydrate
Crystalline Powder
White/Yellow
C16H19N3O5S
cephalexin hydrate, cephalexin monohydrate, cefalexin monohydrate, aristosporin, cefalexgobens, cephalobene, adcadina, azabort, bactopenor, cefacet
O.CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=CC=C1)C2=O)C(O)=O
365.40
CHEBI:3535
≥98.0% (HPLC,T)
Cephalexin Monohydrate
23325-78-2
25 g
MFCD00167148,MFCD00056877
AVGYWQBCYZHHPN-CYJZLJNKSA-N
(6R,7R)-7-[(2R)-2-amino-2-phenylacetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate
62921
347.39

Chemical Identifiers

23325-78-2
365.40
AVGYWQBCYZHHPN-CYJZLJNKSA-N
62921
(6R,7R)-7-[(2R)-2-amino-2-phenylacetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate
C16H19N3O5S
MFCD00167148,MFCD00056877
cephalexin hydrate, cephalexin monohydrate, cefalexin monohydrate, aristosporin, cefalexgobens, cephalobene, adcadina, azabort, bactopenor, cefacet
CHEBI:3535
O.CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=CC=C1)C2=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : XI0350000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions