accessibility menu, dialog, popup

UserName

CAS RN 23325-78-2

IUPAC Name: (6R,7R)-7-[(2R)-2-amino-2-phenylacetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate
Synonyms: cephalexin monohydrate cephalexin.H2O cefalexin hydrate cefalexin monohydrate cefalexin.H2O
Molecular Weight (g/mol): 365.4
Molecular Formula: C16H19N3O5S
InChi Key: AVGYWQBCYZHHPN-CYJZLJNKSA-N
SMILES: O.CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=CC=C1)C2=O)C(O)=O
Color 
  • (2)
  • (2)
Grade 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)